Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
BUFFER BORIC ACID BA4, 40G/L=4% 1 * 1 L

BUFFER BORIC ACID BA4, 40G/L=4% 1 * 1 L

Catalog # B28002
Supplier:   VWR Chemicals
BUFFER BORIC ACID BA4, 40G/L=4% 1 * 1 L
Catalog # B28002
Supplier:  VWR Chemicals
BUFFER BORIC ACID BA4, 40G/L=4% 1 * 1 L
BUFFER BORIC ACID BA4, 40G/L=4% 1 * 1 L
Price
1.180,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.