Bariumnitrat, pure
Glass bottle, 500 g
Leverandør: Thermo Fisher Scientific
Bariumnitrat, pure
Katalog # ACRO453805000
Leverandør: Thermo Fisher Scientific
CAS nummer: 10022-31-8
Gå til:
Product Details & Documents
- CAS nummer:10022-31-8
- Formel:Ba(NO₃)₂
- MDL nummer:MFCD00003442
- Molecular Weight:261.34 g/mol
- Smeltende Pt:592 °C
- Tæthed:~ 3,23 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- FN:1446
- ADR:5.1,II
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g



