Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat, pure

Bariumnitrat, pure

Plastic drum, 10 kg
Katalog # ACRO453800100
Giftig
Oxiderende
Leverandør:   Thermo Fisher Scientific
Bariumnitrat, pure
Katalog # ACRO453800100
CAS nummer:  10022-31-8
Bariumnitrat, pure
Bariumnitrat, pure

Plastic drum, 10 kg

CAS nummer:  10022-31-8
Price
3.050,00 kr
Log ind for tilgængelighed
1 * 10 kg
Product Details & Documents
  • CAS nummer:10022-31-8
  • Formel:Ba(NO₃)₂
  • MDL nummer:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Smeltende Pt:592 °C
  • Tæthed:~ 3,23 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • FN:1446
  • ADR:5.1,II

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder or crystals
  • Titration Complexometric:
    ≥98.5 %

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    10 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.