Propidiumiodid 95%
Glass bottle, 25 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid
- CAS Number:25535-16-4
- Formula:C₂₇H₃₄I₂N₄
- MDL Number:MFCD00011921
- Molecular Weight:668.4 g/mol
- Melting Pt:220…225 °C (dec.)
- Storage Temperature:Køleskab
Documents
ACRO44030
SDS
Specifications
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Melting point:200 to 225 °C
- Titration Argentometric:≥94 %
- Water:≤5 %
Specifications
- Pakning:Glass bottle
- Pakn:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



