Nitrobenzen 99.5% for analysis
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Nitrobenzen 99.5% for analysis
Catalog # ACRO447652500
Supplier: Thermo Fisher Scientific
CAS Number: 98-95-3
Gå til:
Product Details & Documents
- CAS nummer:98-95-3
- Formel:C₆H₅NO₂
- MDL nummer:MFCD00007043
- Molecular Weight:123.11 g/mol
- Kogende Pt:210…211 °C (760 mmHg)
- Smeltende Pt:5…6 °C
- Tæthed:1,205 g/cm³
- Flash Pt:88 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1662
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5520 to 1.5540 (20°C, 589 nm)
- Water:≤0.15 %
- GC:≥99.4 %
- Residue after evaporation:≤0.005 %
- Chloride (Cl):≤5 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



