Nitrobenzen 99.5% for analysis
Glass bottle, 1 L
- :98-95-3
- :C₆H₅NO₂
- :MFCD00007043
- :123.11 g/mol
- :210…211 °C (760 mmHg)
- :5…6 °C
- :1,205 g/cm³
- :88 °C
- :Stue temperatur
- :1662
- :6.1,II
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5520 to 1.5540 (20°C, 589 nm)
- Water:≤0.15 %
- GC:≥99.4 %
- Residue after evaporation:≤0.005 %
- Chloride (Cl):≤5 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Pakning:Glass bottle
- Pakn:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



