Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumthiosulfat, anhydrous 99% for analysis

Natriumthiosulfat, anhydrous 99% for analysis

Plastic bottle, 1 kg
Katalog # ACRO450620010
Leverandør:   Thermo Fisher Scientific
Natriumthiosulfat, anhydrous 99% for analysis
Katalog # ACRO450620010
CAS nummer:  7772-98-7
Natriumthiosulfat, anhydrous 99% for analysis
Natriumthiosulfat, anhydrous 99% for analysis

Plastic bottle, 1 kg

CAS nummer:  7772-98-7
Price
409,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS nummer:7772-98-7
  • Formel:Na₂S₂O₃
  • MDL nummer:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Kogende Pt:100 °C (1013 hPa)
  • Smeltende Pt:48 °C
  • Tæthed:1,66 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Titration Iodimetric:
    ≥98.5 %
  • Loss on drying:
    ≤1 % (105°C)
  • pH:
    6.0 to 8.4 (5 % in water)
  • Chloride (Cl):
    ≤0.05 %
  • Sulfide (S):
    ≤50 ppm
  • Cadmium (Cd):
    ≤50 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Cobalt (Co):
    ≤50 ppm
  • Copper (Cu):
    ≤50 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Nickel (Ni):
    ≤50 ppm
  • Potassium (K):
    ≤100 ppm
  • Zinc (Zn):
    ≤50 ppm
  • Insoluble matter:
    ≤0.01 % (in water)

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifikationer