Brom 99.5+% ACS
Glass bottle, plastic-coated, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:7726-95-6
- Formel:Br₂
- MDL nummer:MFCD00010896
- Molecular Weight:159.81 g/mol
- Kogende Pt:58,7 °C (760 mmHg)
- Smeltende Pt:–7,2 °C
- Tæthed:3,111 g/cm³
- Opbevaringstemperatur:Stue temperatur
- FN:1744
- ADR:8,I
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Dark red-brown Liquid
- Titration Iodometric:≥99.5 %
- Residue after evaporation:≤0.005 %
- Chlorine (Cl):≤0.05 %
- Iodine (I):≤0.001 %
- Nickel (Ni):≤5 ppm
- Heavy metals (as Pb):≤2 ppm
- Sulfur compounds (as S):≤0.001 %
Specifikationer
- Pakning:Glass bottle, plastic-coated
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



