Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Prednison 98%

Glass bottle, 25 g
Katalog # ACRO448950250
Sundhed
Leverandør:   Thermo Fisher Scientific
Prednison 98%
Katalog # ACRO448950250
CAS nummer:  53-03-2
Prednison 98%
Prednison 98%

Glass bottle, 25 g

CAS nummer:  53-03-2
Price
5.810,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:53-03-2
  • Formel:C₂₁H₂₆O₅
  • MDL nummer:MFCD00003608
  • Molecular Weight:358.43 g/mol
  • Smeltende Pt:225…229 °C
  • Opbevaringstemperatur:Stue temperatur

 

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +167° to +175° (20°C, 589 nm) (c= 0.5, dioxane)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.