Prednison 98%
Glass bottle, 5 g
- :53-03-2
- :C₂₁H₂₆O₅
- :MFCD00003608
- :358.43 g/mol
- :225…229 °C
- :Stue temperatur
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥97.5 %
- Specific optical rotation:+167° to +175° (20°C, 589 nm) (c= 0.5, dioxane)
- Pakning:Glass bottle
- Pakn:5 g



