Ethylenglycol, Technical Grade
Plastic drum, 5 L
Leverandør: Thermo Fisher Scientific
Ethylenglycol, Technical Grade
Katalog # ACRO444230050
Leverandør: Thermo Fisher Scientific
CAS nummer: 107-21-1
Gå til:
Product Details & Documents
Synonymer: 1,2-Ethandiol, Glykol
- CAS nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL nummer:MFCD00002885
- Molecular Weight:62.07 g/mol
- Kogende Pt:196…198 °C (760 mmHg)
- Smeltende Pt:–13 °C
- Tæthed:1,113 g/cm³
- Flash Pt:116 °C
- Opbevaringstemperatur:Stue temperatur
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Viscous liquid
- Refractive index:1.4300 to 1.4320 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
Specifikationer
- Pakning:Plastic drum
- Pakn:5 L



