Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyre 99.7+% ACS

Eddikesyre 99.7+% ACS

Plastic drum, 10 L
Catalog # 423220100.
Corrosive
Flammable
Eddikesyre 99.7+% ACS
Catalog # 423220100.
CAS Number:  64-19-7
Eddikesyre 99.7+% ACS
Eddikesyre 99.7+% ACS

Plastic drum, 10 L

CAS Number:  64-19-7
Price
2.940,00 kr
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Boiling Pt:118 °C (1013 hPa)
  • Melting Pt:16…16,5 °C
  • Density:1,048 g/cm³
  • Flash Pt:40 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-580-7
  • UN:2789
  • ADR:8,II
  • REACH:01-2119475328-30

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • GC:
    ≥99.70 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Dilution test:
    Passes test
  • Chloride (Cl):
    ≤1 ppm
  • Sulfate (SO4):
    ≤1 ppm
  • Iron (Fe):
    ≤0.2 ppm
  • Heavy metals (as Pb):
    ≤0.5 ppm
  • Titrable base:
    ≤0.0004 meq/g
  • KMnO4-reducing substance:
    Passes test
  • K2Cr2O7-reducing substance:
    Passes test

Specifications

  • Pakning:
    Plastic drum
  • Pakn:
    10 L

Specifications