Phenol rød ACS
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS nummer:143-74-8
- Formel:C₁₉H₁₄O₅S
- MDL nummer:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Smeltende Pt:>300 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-609-7
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 6.8 (yellow) to pH 8.2 (red)
- Clarity of solution:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



