Phenolphthalein ACS
Plastic bottle, 2,5 kg
: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- :77-09-8
- :C₂₀H₁₄O₄
- :MFCD00005913
- :318.33 g/mol
- :14,07243
- :258…263 °C
- :1,3 g/cm³ (20 °C)
- :Stue temperatur
- :201-004-7
- :01-2119498295-24
- Appearance:Off-white Crystalline powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 8.0 (colorless) to pH 10 (red)
- Clarity of alcohol solution:Passes test
- Pakning:Plastic bottle
- Pakn:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



