Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Tris acetate 99%

Plastic bottle, 500 g
Katalog # ACRO434505000
Leverandør:   Thermo Fisher Scientific
Tris acetate 99%
Katalog # ACRO434505000
CAS nummer:  6850-28-8
Tris acetate 99%
Tris acetate 99%

Plastic bottle, 500 g

CAS nummer:  6850-28-8
Price
5.040,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
  • CAS nummer:6850-28-8
  • Formel:NH₂C(CH₂OH)₃·CH₃COOH
  • MDL nummer:MFCD00038951
  • Molecular Weight:181.1870 g/mol
  • Opbevaringstemperatur:Stue temperatur

 

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Water:
    ≤1.0 %
  • Solubility:
    (1 g/ml in water) Clear colorless
  • UV:
    (50 % in water)
  • Titration with NaOH:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤5 ppm

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    500 g

Specifikationer