Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tris acetate 99%

Tris acetate 99%

Plastic bottle, 100 g
Katalog # ACRO434501000
Leverandør:   Thermo Fisher Scientific
Tris acetate 99%
Katalog # ACRO434501000
CAS nummer:  6850-28-8
Tris acetate 99%
Tris acetate 99%

Plastic bottle, 100 g

CAS nummer:  6850-28-8
Price
1.640,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
  • CAS nummer:6850-28-8
  • Formel:NH₂C(CH₂OH)₃·CH₃COOH
  • MDL nummer:MFCD00038951
  • Molecular Weight:181.1870 g/mol
  • Opbevaringstemperatur:Stue temperatur

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Water:
    ≤1.0 %
  • Solubility:
    (1 g/ml in water) Clear colorless
  • UV:
    (50 % in water)
  • Titration with NaOH:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤5 ppm

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.