Tris acetate 99%
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Stue temperatur
Documents
ACRO43450
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Water:≤1.0 %
- Solubility:(1 g/ml in water) Clear colorless
- UV:(50 % in water)
- Titration with NaOH:≥98.5 %
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pakning:Plastic bottle
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



