Vand ACS
Plastic bottle, 5 L
Supplier: Thermo Fisher Scientific
Vand ACS
Catalog # ACRO434400050
Supplier: Thermo Fisher Scientific
CAS Number: 7732-18-5
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Boiling Pt:100 °C (760 mmHg)
- Melting Pt:0 °C
- Density:1 g/cm³
- Storage Temperature:Stue temperatur
Documents
ACRO43440
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Chloride (Cl):≤0.4 ppm
- Nitrate (NO3):≤0.4 ppm
- Phosphate (PO4):≤1.0 ppm
- Sulfate (SO4):≤1.0 ppm
- Silicate (as SiO2):≤0.01 ppm
- Heavy metals (as Pb):≤0.01 ppm
- KMnO4-reducing substance:Passes test
- Conductivity:2.0 x 10-6 ohm-1 cm-1
Specifications
- Pakning:Plastic bottle
- Pakn:5 L
- Cat. No.:ACRO434400050
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



