Vand ACS
Plastic bottle, 1 L
Vand ACS
ACRO434400010
: 7732-18-5
: Hydrogen oxide
- :7732-18-5
- :H₂O
- :MFCD00011332
- :18.02 g/mol
- :100 °C (760 mmHg)
- :0 °C
- :1 g/cm³
- :Stue temperatur
- Appearance:Clear colorless Liquid
- Chloride (Cl):≤0.4 ppm
- Nitrate (NO3):≤0.4 ppm
- Phosphate (PO4):≤1.0 ppm
- Sulfate (SO4):≤1.0 ppm
- Silicate (as SiO2):≤0.01 ppm
- Heavy metals (as Pb):≤0.01 ppm
- KMnO4-reducing substance:Passes test
- Conductivity:2.0 x 10-6 ohm-1 cm-1
- Pakning:Plastic bottle
- Pakn:1 L
- Cat. No.:ACRO434400010
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



