6-Bromisoquinolin 97%
Glass bottle, 5 g
Leverandør: Thermo Fisher Scientific
6-Bromisoquinolin 97%
Katalog # ACRO429460050
Leverandør: Thermo Fisher Scientific
CAS nummer: 34784-05-9
Gå til:
Product Details & Documents
- CAS nummer:34784-05-9
- Formel:C₉H₆BrN
- MDL nummer:MFCD04973299
- Molecular Weight:208.06 g/mol
- Smeltende Pt:42,6…43.6 °C
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationstestresultater
- Appearance:Orange to light brown Low melting solid
- Infrared spectrum:Conforms
- Melting point:40 to 45 °C
- HPLC:≥96.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



