6-Bromisoquinolin 97%
Glass bottle, 1 g
Leverandør: Thermo Fisher Scientific
6-Bromisoquinolin 97%
Katalog # ACRO429460010
Leverandør: Thermo Fisher Scientific
CAS nummer: 34784-05-9
Gå til:
Product Details & Documents
- CAS nummer:34784-05-9
- Formel:C₉H₆BrN
- MDL nummer:MFCD04973299
- Molecular Weight:208.06 g/mol
- Smeltende Pt:42,6…43.6 °C
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationstestresultater
- Appearance:Orange to light brown Low melting solid
- Infrared spectrum:Conforms
- Melting point:40 to 45 °C
- HPLC:≥96.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



