1,3,2-Dioxathiolane-2,2-dioxide 96%
Glass bottle, 5 g
Leverandør: Thermo Fisher Scientific
1,3,2-Dioxathiolane-2,2-dioxide 96%
Katalog # ACRO426630050
Leverandør: Thermo Fisher Scientific
CAS nummer: 1072-53-3
Gå til:
Product Details & Documents
Appearance: White to beige Powder
Synonymer: 1,3,2-Dioxathiolane 2,2-dioxide, Ethylene Sulphate
- CAS nummer:1072-53-3
- Formel:C₂H₄O₄S
- MDL nummer:MFCD00221769
- Molecular Weight:124.11 g/mol
- Smeltende Pt:95…97 °C
- Opbevaringstemperatur:Køleskab
Documents
ACRO42663
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:95 to 99 °C
- GC:≥95.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



