Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Pentan 95%, Technical Grade

n-Pentan 95%, Technical Grade

Metal drum, 5 L
Katalog # ACRO434030050
Irriterende
Sundhed
Miljø
Brandfarlig
Leverandør:   Thermo Fisher Scientific
n-Pentan 95%, Technical Grade
Katalog # ACRO434030050
CAS nummer:  109-66-0
n-Pentan 95%, Technical Grade
n-Pentan 95%, Technical Grade

Metal drum, 5 L

CAS nummer:  109-66-0
Price
1.340,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
Synonymer:  Pentan
  • CAS nummer:109-66-0
  • Formel:H₃C(CH₂)₃CH₃
  • MDL nummer:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Kogende Pt:36 °C (760 mmHg)
  • Smeltende Pt:–130 °C
  • Tæthed:0,626 g/cm³
  • Flash Pt:–49 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1265
  • ADR:3,II

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3570 to 1.3590 (20°C, 589 nm)
  • GC:
    ≥94 %
  • UV:
    at 265 nm A: ≤1.0
  • Residue after evaporation:
    ≤0.001 %

Specifikationer

  • Pakning:
    Metal drum
  • Pakn:
    5 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.