Methylcyclohexan 99% for spectroscopy
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Methylcyclohexan 99% for spectroscopy
Katalog # ACRO433770010
Leverandør: Thermo Fisher Scientific
CAS nummer: 108-87-2
Gå til:
Product Details & Documents
- CAS nummer:108-87-2
- Formel:C₆H₁₁CH₃
- MDL nummer:MFCD00001497
- Molecular Weight:98.19 g/mol
- Kogende Pt:101 °C (760 mmHg)
- Smeltende Pt:–126,3 °C
- Tæthed:0,77 g/cm³
- Flash Pt:–3 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2296
- ADR:3,II
Documents
ACRO43377
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.5 %
- GC:≥98.5 %
- UV:at 230 nm A: ≤0.15
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



