Imidazol ACS
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS nummer:288-32-4
- Formel:C₃H₄N₂
- MDL nummer:MFCD00005183
- Molecular Weight:68.08 g/mol
- Kogende Pt:255…256 °C (760 mmHg)
- Smeltende Pt:86…90 °C
- Tæthed:1,18 g/cm³ (20 °C)
- Flash Pt:145 °C
- Opbevaringstemperatur:Stue temperatur
- FN:3263
- ADR:8,III
Documents
ACRO39674
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Water:≤0.2 %
- Residue after ignition:≤0.1 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g



