Imidazol ACS
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS nummer:288-32-4
- Formel:C₃H₄N₂
- MDL nummer:MFCD00005183
- Molecular Weight:68.08 g/mol
- Kogende Pt:255…256 °C (760 mmHg)
- Smeltende Pt:86…90 °C
- Tæthed:1,18 g/cm³ (20 °C)
- Flash Pt:145 °C
- Opbevaringstemperatur:Stue temperatur
- FN:3263
- ADR:8,III
Documents
ACRO39674
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Water:≤0.2 %
- Residue after ignition:≤0.1 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



