Imidazol ACS
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS Number:288-32-4
- Formula:C₃H₄N₂
- MDL Number:MFCD00005183
- Molecular Weight:68.08 g/mol
- Boiling Pt:255…256 °C (760 mmHg)
- Melting Pt:86…90 °C
- Density:1,18 g/cm³ (20 °C)
- Flash Pt:145 °C
- Storage Temperature:Stue temperatur
- UN:3263
- ADR:8,III
Specifications
Specification Test Results
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Water:≤0.2 %
- Residue after ignition:≤0.1 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specifications
- Pakning:Plastic bottle
- Pakn:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



