Methylcyclohexan, extra dry 98+%, AcroSeal®
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
Methylcyclohexan, extra dry 98+%, AcroSeal®
Katalog # 397241000.
Leverandør: Thermo Fisher Scientific
CAS nummer: 108-87-2
Gå til:
Product Details & Documents
- CAS nummer:108-87-2
- Formel:C₆H₁₁CH₃
- MDL nummer:MFCD00001497
- Molecular Weight:98.19 g/mol
- Kogende Pt:101 °C (760 mmHg)
- Smeltende Pt:–126,3 °C
- Tæthed:0,77 g/cm³
- Flash Pt:–3 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2296
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.0 %
- Residue after evaporation:≤0.001 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



