Methylcyclohexan, extra dry 98+%, AcroSeal®
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Methylcyclohexan, extra dry 98+%, AcroSeal®
Catalog # 397240010.
Supplier: Thermo Fisher Scientific
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Boiling Pt:101 °C (760 mmHg)
- Melting Pt:–126,3 °C
- Density:0,77 g/cm³
- Flash Pt:–3 °C
- Storage Temperature:Stue temperatur
- UN:2296
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.0 %
- Residue after evaporation:≤0.001 %
Specifications
- Pakning:Glass bottle
- Pakn:1 L



