Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eosin B, pure, certified

Eosin B, pure, certified

Glass bottle, 100 g
Katalog # ACRO409421000
Ætsende
Leverandør:   Thermo Fisher Scientific
Eosin B, pure, certified
Katalog # ACRO409421000
CAS nummer:  548-24-3
Eosin B, pure, certified
Eosin B, pure, certified

Glass bottle, 100 g

CAS nummer:  548-24-3
Price
802,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
  • CAS nummer:548-24-3
  • Formel:C₂₀H₆Br₂N₂Na₂O₉
  • MDL nummer:MFCD00005041
  • Molecular Weight:624.06 g/mol
  • Merck Index:12,03638
  • Smeltende Pt:>310 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-943-1

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark brown to dark red-purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥85 %
  • UV Lambda max:
    511 to 520 nm (in water + 1% Na2CO3)
  • Absorption Ratio P-15/P+15:
    0.96 to 1.22
  • BSC Certification:
    Passes

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.