Ammoniumchlorid for analysis
Glass bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Ammoniumchlorid for analysis
Katalog # ACRO393180010
Leverandør: Thermo Fisher Scientific
CAS nummer: 12125-02-9
Gå til:
Product Details & Documents
Synonymer: Ammonium muriate, Salmiac
- CAS nummer:12125-02-9
- Formel:NH₄Cl
- MDL nummer:MFCD00011420
- Molecular Weight:53.49 g/mol
- Smeltende Pt:340 °C (subl.)
- Tæthed:1,5256 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99.0 %
- Residue after ign. (sulfated):≤0.01 %
- Phosphate (PO4):≤0.002 %
- Sulfate (SO4):≤0.002 %
- Calcium (Ca):≤0.001 %
- Iron (Fe):≤0.0005 %
- Magnesium (Mg):≤0.001 %
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 % (5 % in water)
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg



