Geraniol 99%
Plastic drum, 2,5 kg
: (E)-3,7-Dimethylocta-2,6-dien-1-ol
- :106-24-1
- :C₁₀H₁₈O
- :MFCD00002917
- :154.25 g/mol
- :229…230 °C (757 mmHg)
- :–15 °C
- :0,878 g/cm³
- :76 °C
- :Stue temperatur
- :203-377-1
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4780 (20°C, 589 nm)
- GC:≥98.5 %
- Pakning:Plastic drum
- Pakn:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



