Methylethylketon ACS
Plastic drum, 10 L
Methylethylketon ACS
ACRO389430100
: 78-93-3
: Ethylmethylketon, Butanon
- :78-93-3
- :C₂H₅COCH₃
- :MFCD00011648
- :72.11 g/mol
- :13,06097
- :80 °C (760 mmHg)
- :–87 °C
- :0,806 g/cm³
- :–7 °C
- :Stue temperatur
- :201-159-0
- :1193
- :3,II
- :01-2119457290-43
- Appearance:Clear liquid
- Water:≤0.20 %
- GC:≥99.0 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.0025 %
- Titrable acid:≤0.0005 meq/g
- Pakning:Plastic drum
- Pakn:10 L
- Cat. No.:ACRO389430100
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



