Lithiumchlorid 99+% ACS
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Lithiumchlorid 99+% ACS
Katalog # ACRO413271000
Leverandør: Thermo Fisher Scientific
CAS nummer: 7447-41-8
Gå til:
Product Details & Documents
- CAS nummer:7447-41-8
- Formel:LiCl
- MDL nummer:MFCD00011078
- Molecular Weight:42.39 g/mol
- Merck Index:13,05550
- Kogende Pt:1382 °C (760 mmHg)
- Smeltende Pt:605 °C
- Tæthed:2,06 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-212-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99 %
- Loss on drying:≤1.0 % (1 g, 105°C, 2 hrs)
- Sulfate (SO4):≤100 ppm
- Barium (Ba):≤30 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤2000 ppm
- Heavy metals (ICP-OES):≤20 ppm
- Titrable base:≤0.008 meq/g
- Insoluble matter:≤0.01 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g



