Cellulose, microcrystalline ∼ Ø 50 µm
Plastic bottle, 250 g
Leverandør: Thermo Fisher Scientific
Cellulose, microcrystalline ∼ Ø 50 µm
Katalog # ACRO387242500
Leverandør: Thermo Fisher Scientific
CAS nummer: 9004-34-6
Gå til:
Product Details & Documents
Synonymer: Microcrystallinecellulose
- CAS nummer:9004-34-6
- Formel:(C₁₂H₂₂O₁₁)n
- MDL nummer:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:13,01977
- Smeltende Pt:260 - 270 °C
- Tæthed:1,27 to 1.6 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-674-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1.5 %
- Particle size:≤1 % (> 250 µm)
- Residue after ignition:≤0.05 %
- pH:5 to 7
- Heavy metals (as Pb):≤10 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



