Natriumoctadecylsulfat 98%
Glass bottle, 5 g
Leverandør: Thermo Fisher Scientific
Natriumoctadecylsulfat 98%
Katalog # ACRO384890050
Leverandør: Thermo Fisher Scientific
CAS nummer: 1120-04-3
Gå til:
Product Details & Documents
Synonymer: Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt
- CAS nummer:1120-04-3
- Formel:C₁₈H₃₇NaO₄S
- MDL nummer:MFCD00007469
- Molecular Weight:372.55 g/mol
- Smeltende Pt:211…213 °C
- Opbevaringstemperatur:Stue temperatur
Documents
ACRO38489
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow to light beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤7 %
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



