Natriumoctadecylsulfat 98%
Glass bottle, 1 g
: Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt
- :1120-04-3
- :C₁₈H₃₇NaO₄S
- :MFCD00007469
- :372.55 g/mol
- :211…213 °C
- :Stue temperatur
- Appearance:White to light yellow to light beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤7 %
- GC:≥97.5 %
- Pakning:Glass bottle
- Pakn:1 g



