Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromphenol blå ACS

Bromphenol blå ACS

Glass bottle, 50 g
Catalog # ACRO403140500
Bromphenol blå ACS
Catalog # ACRO403140500
CAS Number:  115-39-9
Bromphenol blå ACS
Bromphenol blå ACS

Glass bottle, 50 g

CAS Number:  115-39-9
Price
2.220,00 kr
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:279 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:204-086-2

Documents

Specifications

Specification Test Results

  • Appearance:
    Pink-orange to red to purple Powder
  • Infrared spectrum:
    Conforms
  • Visual transition interval:
    from pH 3.0 (yellow) to pH 4.6 (blue)
  • Clarity of solution:
    Passes test

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    50 g

Specifications