2-Propanol 99.5%, gradient grade for HPLC
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
2-Propanol 99.5%, gradient grade for HPLC
Catalog # ACRO383920010
Supplier: Thermo Fisher Scientific
CAS Number: 67-63-0
Jump to:
Product Details & Documents
Synonyms: Isopropylalkohol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:81…83 °C (760 mmHg)
- Melting Pt:–89,5 °C
- Density:0,785 g/cm³
- Flash Pt:12 °C
- Storage Temperature:Stue temperatur
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.1 %
- GC:≥99.4 %
- UV:at 260 nm A: ≤0.005
- Residue after evaporation:≤2 ppm
- Acidity/alkalinity:≤0.0001 meq/g
- Gradient analysis::
- Absorbance largest eluted peak:≤0.002 A.U.
- Filtered to 0.2 micrometer:Filtered
Specifications
- Pakning:Glass bottle
- Pakn:1 L
- Cat. No.:ACRO383920010
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



