Triethylcitrat 99%
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- CAS nummer:77-93-0
- Formel:C₁₂H₂₀O₇
- MDL nummer:MFCD00009201
- Molecular Weight:276.29 g/mol
- Kogende Pt:294 °C (760 mmHg)
- Smeltende Pt:–46 °C
- Tæthed:1,136 g/cm³
- Flash Pt:151 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-070-7
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4430 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤50 APHA
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg



