Isooctan 99.5% for analysis
Metal drum, 25 L
Leverandør: Thermo Fisher Scientific
Isooctan 99.5% for analysis
Katalog # ACRO265440250
Leverandør: Thermo Fisher Scientific
CAS nummer: 540-84-1
Gå til:
Product Details & Documents
Synonymer: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- CAS nummer:540-84-1
- Formel:(CH₃)₂CHCH₂C(CH₃)₃
- MDL nummer:MFCD00008943
- Molecular Weight:114.23 g/mol
- Merck Index:13,05212
- Kogende Pt:98…99 °C (760 mmHg)
- Smeltende Pt:–107 °C
- Tæthed:0,69 g/cm³
- Flash Pt:–12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-759-1
- FN:1262
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.01 % (Coulometric)
- GC:≥99.4 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Aluminium (Al):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Nickel (Ni):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- H2S+foreign org. S cmpds:≤50 ppm
Specifikationer
- Pakning:Metal drum
- Pakn:25 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



