2'-Fluor-4'-methoxyacetophenon 99%
Glass bottle, 25 g
- :74457-86-6
- :C₉H₉FO₂
- :MFCD00042290
- :168.17
- :80…85 °C (,1 mmHg)
- :50…54 °C
- :>110 °C
- :Stue temperatur
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:50 to 54 °C
- GC:≥98.5 %
- Pakning:Glass bottle
- Pakn:25 g



