1-Octen 99+%
Glass bottle, 500 mL
- :111-66-0
- :C₈H₁₆
- :MFCD00009548
- :112.22 g/mol
- :13,01770
- :121…123 °C (760 mmHg)
- :–102 °C
- :0,715 g/cm³
- :13 °C
- :Stue temperatur
- :203-893-7
- :3295
- :3,II
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4080 to 1.4100 (20°C, 589 nm)
- Water:≤0.005 %
- GC:≥99.0 %
- Pakning:Glass bottle
- Pakn:500 mL



