Behensyre 85%, Technical Grade
Glass bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Behensyre 85%, Technical Grade
Katalog # ACRO334810010
Leverandør: Thermo Fisher Scientific
CAS nummer: 112-85-6
Gå til:
Product Details & Documents
Synonymer: Docosanoic acid
- CAS nummer:112-85-6
- Formel:C₂₂H₄₄O₂
- MDL nummer:MFCD00002807
- Molecular Weight:340.59 g/mol
- Merck Index:13,01024
- Kogende Pt:306 °C (60 mmHg)
- Smeltende Pt:80…82 °C
- Tæthed:0,8221 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-010-8
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- GC:≥80 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg



