Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexan, extra dry 96+%, AcroSeal®

n-Hexan, extra dry 96+%, AcroSeal®

Glass bottle, 2,5 L
Katalog # 326920025.
Irriterende
Sundhed
Miljø
Brandfarlig
Leverandør:   Thermo Fisher Scientific
n-Hexan, extra dry 96+%, AcroSeal®
Katalog # 326920025.
CAS nummer:  110-54-3
n-Hexan, extra dry 96+%, AcroSeal®
n-Hexan, extra dry 96+%, AcroSeal®

Glass bottle, 2,5 L

CAS nummer:  110-54-3
Price
2.770,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Kogende Pt:69 °C (760 mmHg)
  • Smeltende Pt:–95 °C
  • Tæthed:0,659 g/cm³
  • Flash Pt:–22 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-777-6
  • FN:1208
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.002 %
  • GC:
    ≥96.0 %
  • Residue after evaporation:
    ≤0.0005 %
  • Filt.over 0.2 micron PFTE:
    Filtered

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.