Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cyclohexan, extra dry 99.5%, AcroSeal®

Glass bottle, AcroSeal®, 1 L
Katalog # 326830010.
Irriterende
Sundhed
Miljø
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Cyclohexan, extra dry 99.5%, AcroSeal®
Katalog # 326830010.
CAS nummer:  110-82-7
Cyclohexan, extra dry 99.5%, AcroSeal®
Cyclohexan, extra dry 99.5%, AcroSeal®

Glass bottle, AcroSeal®, 1 L

CAS nummer:  110-82-7
Price
911,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Hexahydrobenzene, Hexamethylene
  • CAS nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL nummer:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Kogende Pt:81 °C (760 mmHg)
  • Smeltende Pt:6,5 °C
  • Tæthed:0,77 g/cm³
  • Flash Pt:–18 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-806-2
  • FN:1145
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.4 %
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.001 meq/g

Specifikationer

  • Pakning:
    Glass bottle, AcroSeal®
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.