MES, (max. 1% H₂O) 99+%
Glass bottle, 250 g
Leverandør: Thermo Fisher Scientific
MES, (max. 1% H₂O) 99+%
Katalog # ACRO274142500
Leverandør: Thermo Fisher Scientific
CAS nummer: 4432-31-9
Gå til:
Product Details & Documents
Synonymer: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
- CAS nummer:4432-31-9
- Formel:C₆H₁₃NO₄S
- MDL nummer:MFCD00006181
- Molecular Weight:195.24 g/mol
- Smeltende Pt:>300 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
Documents
ACRO27414
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- Solubility:(0.5 M in water) Clear colorless
- Titration with NaOH:≥99.0 % (On anhydrous basis)
Specifikationer
- Pakning:Glass bottle
- Pakn:250 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



