MES, (max. 1% H₂O) 99+%
Glass bottle, 1 kg
: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
- :4432-31-9
- :C₆H₁₃NO₄S
- :MFCD00006181
- :195.24 g/mol
- :>300 °C (dec.)
- :Stue temperatur
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- Solubility:(0.5 M in water) Clear colorless
- Titration with NaOH:≥99.0 % (On anhydrous basis)
- Pakning:Glass bottle
- Pakn:1 kg
- Cat. No.:ACRO274140010



