Procain, HCl 99%
Plastic bottle, 500 g
: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
- :51-05-8
- :C₁₃H₂₁ClN₂O₂
- :MFCD00013000
- :272.77 g/mol
- :154…158 °C
- :Stue temperatur
- :200-077-2
- :2811
- :6.1,III
- Appearance:Colorless to white to almost white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Sulfated ash:≤0.1 %
- pH:5.0 to 6.5 (2 % w/v in water)
- Titration with NaOH:98.5 to 101.5 %
- Pakning:Plastic bottle
- Pakn:500 g



