Natriumcarbonat 99+%, granular for analysis
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Natriumcarbonat 99+%, granular for analysis
Catalog # ACRO206800050
Supplier: Thermo Fisher Scientific
CAS Number: 497-19-8
Jump to:
Product Details & Documents
Synonyms: Calcined soda, Carbonic acid disodium salt
- CAS Number:497-19-8
- Formula:Na₂CO₃
- MDL Number:MFCD00003494
- Molecular Weight:105.99 g/mol
- Merck Index:14,08596
- Boiling Pt:1600 °C (760 mmHg)
- Melting Pt:851 °C
- Density:2,53 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:207-838-8
- REACH:01-2119485498-19
Documents
ACRO20680
SDS
Specifications
Specification Test Results
- Appearance:White Granular powder
- Loss on drying:≤0.5 % (285°C)
- Titration with HCl:≥99.0 %
- Chloride (Cl):≤20 ppm
- Phosphate (PO4):≤10 ppm
- Arsenic (As):≤1 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤5 ppm
- Potassium (K):≤300 ppm
- Heavy metals (as Pb):≤20 ppm
- Total nitrogen compounds:≤0.002 % (as N)
- Silicon dioxide (SiO2):≤50 ppm
- Total sulfur (as SO4):≤0.005 %
Specifications
- Pakning:Plastic drum
- Pakn:5 kg



